Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028608.3 | NP_082884.1 | 3 | 713 | ||
GenBank mRNA | AK010768.1 | - | 3 | 645 | ||
AK137557.1 | - | 3 | 660 | |||
AK170051.1 | - | 3 | 660 | |||
BC025083.1 | - | 3 | 637 | 3711757 | ||
BC098231.1 | - | 3 | 687 | 3325823 | ||
BY540964.1 | - | 11 |