Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028636.2 | NP_082912.1 | 3, 4 | 360 | ||
GenBank mRNA | AK049370.1 | - | 3, 4 | 381 | ||
AK153935.1 | - | 3, 4 | 381 | |||
AK158387.1 | - | 3, 4 | 367 | |||
AK163077.1 | - | 3, 4 | 367 | |||
AK170642.1 | - | 3, 4 | 390 | |||
AK171110.1 | - | 3, 4 | 370 | |||
AK171599.1 | - | 3, 4 | 357 | |||
BC016253.1 | - | 3, 4 | 360 | 4911979 |