Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028636.2 | NP_082912.1 | 4 | 384 | ||
GenBank mRNA | AK049370.1 | - | 4 | 405 | ||
AK153935.1 | - | 4 | 405 | |||
AK158387.1 | - | 4 | 391 | |||
AK163077.1 | - | 4 | 391 | |||
AK170642.1 | - | 4 | 414 | |||
AK171110.1 | - | 4 | 394 | |||
AK171599.1 | - | 4 | 381 | |||
BC016253.1 | - | 4 | 384 | 4911979 |