Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028636.2 | NP_082912.1 | 7, 8 | 914 | ||
GenBank mRNA | AK049370.1 | - | 7, 8 | 935 | ||
AK153935.1 | - | 7, 8 | 935 | |||
AK163077.1 | - | 7, 8 | 921 | |||
AK171110.1 | - | 8, 9 | 1047 | |||
AK171599.1 | - | 7, 8 | 911 | |||
BC016253.1 | - | 7, 8 | 914 | 4911979 |