Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028665.4 | NP_082941.2 | 3, 4 | 1293 | ||
GenBank mRNA | AK029511.1 | - | 3, 4 | 1295 | ||
AK029888.1 | - | 3, 4 | 1254 | |||
AK033599.1 | - | 3, 4 | 1282 | |||
AK048925.1 | - | 3, 4 | 1187 | |||
AK133339.1 | - | 1254 | ||||
BC030885.1 | - | 3, 4 | 922 | |||
BC100433.1 | - | 3, 4 | 1287 | |||
BC145456.1 | - | 3, 4 | 411 | 9054017 | ||
BC145457.1 | - | 3, 4 | 411 | |||
BC145458.1 | - | 3, 4 | 411 |