Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D3Ertd751e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099785.1 | NP_001093255.1 | 6 | 645 | ||
NM_027271.1 | NP_081547.1 | 5 | 510 | |||
NM_028667.3 | NP_082943.2 | 7 | 762 | |||
GenBank mRNA | AK015146.1 | - | 6 | 716 | ||
AK039725.1 | - | 6 | 609 | |||
AK044548.1 | - | 4 | 348 | |||
AK045813.1 | - | 6 | 609 | |||
AK085404.1 | - | 6 | 643 | |||
AK138405.1 | - | 8 | 867 | |||
AK144064.1 | - | 6 | 609 | |||
BC062244.1 | - | 5 | 470 | 1345984 |