Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006344.2 | NP_006335.2 | 6 | 728 | ||
NM_182906.2 | NP_878910.1 | 6 | 809 | |||
XM_005256412.1 | XP_005256469.1 | 6 | 754 | |||
XM_011523613.1 | XP_011521915.1 | 6 | 835 | |||
XM_011523614.2 | XP_011521916.1 | 7 | 901 | |||
XM_011523615.1 | XP_011521917.1 | 6 | 835 | |||
GenBank mRNA | AK291975.1 | - | 6 | 691 | ||
AK292363.1 | - | 6 | 695 | |||
AL546075.3 | - | 6 | 677 | |||
BC027858.1 | - | 6 | 675 | 5225381 | ||
BC039011.1 | - | 6 | 757 | 5751735 | ||
D50532.1 | - | 6 | 632 |