Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159948.2 | NP_001153420.2 | 4 | 1050 | ||
NM_028717.6 | NP_082993.4 | 4 | 881 | |||
NM_146109.3 | NP_666221.1 | 4 | 881 | |||
GenBank mRNA | AB053307.1 | - | 4 | 869 | ||
AK029858.1 | - | 4 | 1051 | |||
BC031479.1 | - | 4 | 858 | 3257574 | ||
BC046828.1 | - | 4 | 842 | 6409924 |