Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028721.3 | NP_082997.3 | 21 | 3133 | ||
GenBank mRNA | AK014393.1 | - | 6 | 693 | ||
AK133217.1 | - | 21 | 3134 | |||
AK173327.1 | - | 21 | 2712 | |||
AY259499.1 | - | 21 | 3070 | |||
BC115724.1 | - | 21 | 2660 | 40086844 |