Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028722.2 | NP_082998.2 | 2 | 928 | ||
GenBank mRNA | AK014463.1 | - | 2 | 1181 | ||
AK015496.1 | - | 2 | 192 | |||
AK053719.1 | - | 2 | 898 | |||
AK161412.1 | - | 2 | 171 | |||
BC043039.1 | - | 2 | 906 | 5701204 | ||
BC055715.1 | - | 2 | 880 | 6414174 | ||
BC058629.1 | - | 2 | 928 | 6402215 | ||
DQ387855.1 | - | 2 | 191 |