Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014568.1 | - | 1 | 242 | ||
AK042581.1 | - | 1 | 228 | |||
AK046761.1 | - | 1 | 233 | |||
AK047599.1 | - | 1 | 256 | |||
AK050948.1 | - | 1 | 240 | |||
AK157398.1 | - | 1 | 240 | |||
AK164493.1 | - | 1 | 250 | |||
AK172043.1 | - | 1 | 245 | |||
BC024877.1 | - | 1 | 244 | |||
BC051109.1 | - | 1 | 213 | |||
BC096599.1 | - | 1 | 232 | |||
BC125432.1 | - | 1 | 239 | 40129947 | ||
BC132099.1 | - | 1 | 239 | 40130376 |