Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271411.1 | NP_001258340.1 | 2 | 454 | ||
NM_001271412.1 | NP_001258341.1 | 2 | 384 | |||
NM_001271413.1 | NP_001258342.1 | 2 | 293 | |||
NM_001271414.1 | NP_001258343.1 | 2 | 381 | |||
NM_028728.3 | NP_083004.1 | 2 | 311 | |||
GenBank mRNA | AF361364.1 | - | 2 | 388 | ||
AK014802.2 | - | 2 | 312 | |||
AK048992.1 | - | 2 | 295 | |||
AK150635.1 | - | 2 | 363 | |||
AK151187.1 | - | 2 | 363 | |||
AK151884.1 | - | 2 | 363 | |||
AK152262.1 | - | 2 | 363 | |||
AK152499.1 | - | 2 | 363 | |||
AK152688.1 | - | 2 | 363 | |||
AK162939.1 | - | 2 | 365 | |||
AK170355.1 | - | 2 | 361 | |||
AK170442.1 | - | 2 | 363 | |||
AK170545.1 | - | 2 | 249 | |||
AK170593.1 | - | 2 | 319 | |||
AK170677.1 | - | 2 | 319 |