Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006351.3 | NP_006342.2 | 3 | 449 | ||
GenBank mRNA | AF026030.1 | - | 3 | 266 | ||
AF041254.1 | - | 3 | 307 | |||
AK223062.1 | - | 3 | 292 | |||
AK289634.1 | - | 3 | 449 | |||
AK309415.1 | - | 3 | 293 | |||
BC033628.1 | - | 3 | 279 | 5248163 |