Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028761.3 | NP_083037.1 | 7 | 581 | ||
GenBank mRNA | AK004572.1 | - | 7 | 582 | ||
AK041044.1 | - | 7 | 550 | |||
AK045181.1 | - | 7 | 567 | |||
AK049829.1 | - | 5 | 1657 | |||
AK160611.1 | - | 7 | 550 | |||
BC021899.1 | - | 7 | 549 | 4015784 |