Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028780.3 | NP_083056.2 | 2 | 612 | ||
GenBank mRNA | AF269152.1 | - | 2 | 260 | ||
AK004754.2 | - | 2 | 614 | |||
AK090065.1 | - | 1 | 607 | |||
AK152476.1 | - | 2 | 392 | |||
AK152540.1 | - | 2 | 392 | |||
AK160610.1 | - | 1 | 604 | |||
BC017617.1 | - | 1 | 551 | 4923337 |