Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028780.3 | NP_083056.2 | 5 | 1780 | ||
GenBank mRNA | AF269152.1 | - | 6 | 1425 | ||
AK004754.2 | - | 5 | 1782 | |||
AK090065.1 | - | 4 | 1776 | |||
AK152476.1 | - | 5 | 1560 | |||
AK152540.1 | - | 5 | 1560 | |||
BC007187.1 | - | 4 | 1085 | |||
BC017617.1 | - | 4 | 1719 | 4923337 |