Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028792.3 | NP_083068.1 | 3 | 1281 | ||
GenBank mRNA | AK004913.1 | - | 3 | 1230 | ||
AK154799.1 | - | 3 | 1330 | |||
AK214040.1 | - | 2 | 145 | |||
BC006928.1 | - | 3 | 429 | 2655826 | ||
BC086769.1 | - | 3 | 401 | 5707990 |