Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006366.2 | NP_006357.1 | 6 | 757 | ||
XM_011514233.2 | XP_011512535.1 | 5 | 968 | |||
GenBank mRNA | AK297538.1 | - | 5 | 473 | ||
AK308461.1 | - | 5 | 472 | |||
AK312362.1 | - | 6 | 885 | |||
BC008481.1 | - | 6 | 550 | 4295984 | ||
CR457022.1 | - | 5 | 514 | |||
U02390.1 | - | 6 | 597 |