Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028808.3 | NP_083084.2 | 2 | 1282 | ||
GenBank mRNA | AK008013.2 | - | 2 | 1283 | ||
AK038560.1 | - | 2 | 1283 | |||
AK083264.1 | - | 2 | 1283 | |||
BC119043.1 | - | 2 | 1279 | 8733792 |
Set Membership: |
G-protein coupled receptor |