Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253812.1 | NP_001240741.1 | 5 | 523 | ||
NM_028811.3 | NP_083087.1 | 5 | 466 | |||
GenBank mRNA | AK012072.1 | - | 5 | 404 | ||
AK088457.1 | - | 5 | 487 | |||
AK150058.1 | - | 5 | 404 | |||
BC026461.1 | - | 3 | 118 | |||
BC057453.1 | - | 5 | 406 | 3969625 |