Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253812.1 | NP_001240741.1 | 6 | 586 | ||
NM_028811.3 | NP_083087.1 | 6 | 529 | |||
GenBank mRNA | AK012072.1 | - | 6 | 467 | ||
AK088457.1 | - | 6 | 550 | |||
AK150058.1 | - | 6 | 467 | |||
BC026461.1 | - | 4 | 181 | |||
BC048822.1 | - | 1 | 34 | |||
BC057453.1 | - | 6 | 469 | 3969625 |