Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253812.1 | NP_001240741.1 | 15 | 2029 | ||
NM_028811.3 | NP_083087.1 | 15 | 1972 | |||
GenBank mRNA | AK012072.1 | - | 15 | 1910 | ||
AK088457.1 | - | 15 | 1993 | |||
AK150058.1 | - | 15 | 2061 | |||
BC026461.1 | - | 13 | 1624 | |||
BC037470.1 | - | 9 | 1326 | |||
BC048822.1 | - | 10 | 1477 | |||
BC057453.1 | - | 15 | 1912 | 3969625 |