Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028816.2 | NP_083092.2 | 4 | 817 | ||
GenBank mRNA | AK049009.1 | - | 4 | 830 | ||
AK154709.1 | - | 4 | 859 | |||
AY029528.1 | - | 3 | 310 | |||
BC058090.1 | - | 4 | 817 | 6826356 |