Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006369.4 | NP_006360.3 | 4 | 1052 | ||
XM_017000074.1 | XP_016855563.1 | 4 | 1078 | |||
GenBank mRNA | AF370430.1 | - | 4 | 952 | ||
AK024051.1 | - | 4 | 961 | |||
AK125827.1 | - | 3 | 2329 | |||
AK223420.1 | - | 1 | 414 | |||
AK291283.1 | - | 4 | 1035 | |||
AK296436.1 | - | 4 | 871 | |||
AK297596.1 | - | 4 | 1066 | |||
BC004953.1 | - | 1 | 288 | |||
BC092479.1 | - | 4 | 862 | |||
BC101522.1 | - | 4 | 1042 | 8069028 | ||
BC101524.1 | - | 4 | 1042 | 8069030 | ||
BX537366.1 | - | 4 | 962 | |||
X86018.1 | - | 1 | 264 |