Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028846.5 | NP_083122.1 | 21 | 2476 | ||
GenBank mRNA | AF449715.1 | - | 21 | 2427 | ||
AK006800.1 | - | 4 | 410 | |||
AK029890.1 | - | 2372 | ||||
AK047242.1 | - | 21 | 2395 | |||
AK054279.1 | - | 21 | 2419 | |||
AK087412.1 | - | 21 | 2388 | |||
AK163663.1 | - | 21 | 2386 | |||
AK173083.1 | - | 21 | 2378 | |||
BC019735.1 | - | 7 | 676 | |||
BC037199.1 | - | 15 | 1860 | |||
BC079674.1 | - | 2365 | 30531994 |