Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028860.2 | NP_083136.2 | 7 | 800 | ||
GenBank mRNA | AK141138.1 | - | 7 | 780 | ||
BC032166.1 | - | 7 | 783 | 5352919 | ||
BC081544.1 | - | 7 | 784 | 3672099 |