Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028876.2 | NP_083152.1 | 4 | 1750 | ||
GenBank mRNA | AK014490.1 | - | 4 | 1751 | ||
AK033463.1 | - | 4 | 1675 | |||
AK081097.1 | - | 1 | 429 | |||
AK159813.1 | - | 4 | 2810 | |||
AK170036.1 | - | 5 | 1679 |