Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rptor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB082952.1 | - | 4 | 441 | ||
AK016530.1 | - | 4 | 588 | |||
AK029341.1 | - | 4 | 1237 | |||
AK087217.1 | - | 4 | 1257 | |||
AK144082.1 | - | 4 | 1236 | |||
AK161011.1 | - | 4 | 1347 | |||
AK172276.1 | - | 4 | 1234 | |||
BC060140.1 | - | 4 | 553 | |||
BC118971.1 | - | 4 | 436 | |||
BC148309.2 | - | 4 | 441 |