Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Batf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028967.1 | NP_083243.1 | 3 | 762 | ||
GenBank mRNA | AK016990.1 | - | 978 | |||
AK150133.1 | - | 3 | 782 | |||
AK171828.1 | - | 3 | 779 | |||
BC024521.1 | - | 3 | 762 | 4984403 |