Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006391.2 | NP_006382.1 | 2, 3 | 316 | ||
GenBank mRNA | AF098799.1 | - | 2, 3 | 281 | ||
AK312885.1 | - | 2, 3 | 287 | |||
BC114929.1 | - | 2, 3 | 242 | 40036258 | ||
DA780921.1 | - | 2, 3 | 316 |