Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 8430419L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028982.4 | NP_083258.2 | 3 | 554 | ||
GenBank mRNA | AK018428.1 | - | 3 | 498 | ||
AK039401.1 | - | 3 | 490 | |||
AK155128.1 | - | 3 | 498 | |||
AK169892.1 | - | 3 | 555 | |||
AK220395.1 | - | 2 | 204 | |||
BC038002.1 | - | 2 | 215 | |||
BC079676.1 | - | 3 | 481 | |||
BC137674.1 | - | 3 | 533 | 8860693 | ||
BC137676.1 | - | 3 | 533 | 8860695 |