Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 8430419L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028982.4 | NP_083258.2 | 6 | 999 | ||
GenBank mRNA | AK018428.1 | - | 6 | 943 | ||
AK039401.1 | - | 6 | 935 | |||
AK155128.1 | - | 6 | 943 | |||
AK169892.1 | - | 6 | 1000 | |||
AK220395.1 | - | 5 | 649 | |||
BC038002.1 | - | 5 | 660 | |||
BC079676.1 | - | 6 | 926 | |||
BC137674.1 | - | 6 | 978 | 8860693 | ||
BC137676.1 | - | 6 | 978 | 8860695 |