Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029017.2 | NP_083293.1 | 4, 5 | 432 | ||
GenBank mRNA | BC029173.1 | - | 412 | |||
BC038065.1 | - | 4, 5 | 403 | |||
BC087953.1 | - | 4, 5 | 426 | 30119240 | ||
CX122942.1 | - | 4, 5 | 411 |