Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006397.2 | NP_006388.2 | 6 | 668 | ||
XM_006722619.2 | XP_006722682.1 | 7 | 853 | |||
GenBank mRNA | AK315327.1 | - | 6 | 698 | ||
AY363912.1 | - | 6 | 657 | |||
BC011748.2 | - | 6 | 659 | 3160621 | ||
BE560339.1 | - | 14 | ||||
BG825340.1 | - | 6 | 672 | |||
Z97029.1 | - | 6 | 668 |