Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164366.1 | NP_001157838.1 | 3 | 454 | ||
NM_029097.2 | NP_083373.2 | 3 | 454 | |||
GenBank mRNA | AK156104.1 | - | 3 | 434 | ||
AK171305.1 | - | 3 | 407 | |||
AK171554.1 | - | 3 | 458 | |||
AK209090.1 | - | 1 | 65 | |||
BC023746.1 | - | 3 | 429 | 5353766 | ||
BC025028.1 | - | 3 | 349 | |||
BC042661.1 | - | 3 | 399 | 4192094 |