Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006404.4 | NP_006395.2 | 4 | 926 | ||
GenBank mRNA | BC014451.1 | - | 4 | 790 | 4907433 | |
BQ939781.1 | - | 2 | 258 | |||
L35545.1 | - | 4 | 732 | |||
X89079.1 | - | 4 | 737 |