Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014842.2 | NP_001014842.1 | 2 | 584 | ||
NM_006405.6 | NP_006396.2 | 2 | 584 | |||
GenBank mRNA | AK292110.1 | - | 2 | 273 | ||
BC010856.2 | - | 1 | 193 | 3899246 | ||
BM922952.1 | - | 535 | ||||
BX161390.1 | - | 2 | 504 | |||
BX161494.1 | - | 2 | 466 | |||
U94831.1 | - | 1 | 191 |