Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1bp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029166.2 | NP_083442.2 | 2, 3 | 468 | ||
GenBank mRNA | AB093261.1 | - | 2, 3 | 433 | ||
AK033300.1 | - | 3, 4 | 476 | |||
AK141050.1 | - | 2, 3 | 476 | |||
BC030383.1 | - | 427 | ||||
BC132134.1 | - | 2, 3 | 261 | 40130411 | ||
BC138092.1 | - | 2, 3 | 261 | 8861111 |