Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171004.1 | NP_001164475.1 | 3 | 1139 | ||
NM_001171005.1 | NP_001164476.1 | 2 | 335 | |||
NM_029239.3 | NP_083515.2 | 3 | 1139 | |||
GenBank mRNA | AK016167.1 | - | 3 | 294 | ||
AK030058.1 | - | 2 | 332 | |||
AK050059.1 | - | 1 | 163 | |||
BC027320.1 | - | 2 | 540 | |||
BC037012.1 | - | 2 | 783 | 5342428 | ||
BC089164.1 | - | 3 | 1038 | 5400250 |