Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029271.2 | NP_083547.1 | 3 | 375 | ||
GenBank mRNA | AB049650.1 | - | 3 | 406 | ||
AF279255.1 | - | 3 | 352 | |||
AK002753.1 | - | 3 | 376 | |||
AK031791.1 | - | 1 | 2333 | |||
AK078324.1 | - | 3 | 393 | |||
BC037368.1 | - | 2 | 79 | |||
BC052214.1 | - | 3 | 410 | 30018135 |
Gene Symbol: Gm6578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC099426.1 | - | 4 | 585 |