Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198865.1 | NP_942565.1 | 2 | 4599 | ||
GenBank mRNA | BC065107.1 | - | 3 | 3944 | 30357422 |