Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031684.2 | NP_001026854.1 | 8 | 1368 | ||
NM_001195446.1 | NP_001182375.1 | 7 | 1332 | |||
GenBank mRNA | AK091425.1 | - | 9 | 1632 | ||
AK225141.1 | - | 7 | 1145 | |||
AK293667.1 | - | 6 | 1016 | |||
AY513287.1 | - | 8 | 1211 | |||
BC022328.1 | - | 8 | 1233 | 4251314 |