Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029341.1 | NP_083617.2 | 3 | 418 | ||
GenBank mRNA | AK006467.1 | - | 3 | 418 | ||
BC037621.1 | - | 2 | 197 | |||
BC061015.1 | - | 3 | 376 | 30297953 |