Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029385.2 | NP_083661.2 | 3 | 706 | ||
GenBank mRNA | AK009737.1 | - | 3 | 712 | ||
AK012920.1 | - | 2 | 759 | |||
AK013493.1 | - | 1 | 1542 | |||
BC064048.1 | - | 3 | 693 | 574013 |