Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193309.1 | NP_001180238.1 | 2, 3 | 457 | ||
NM_029413.4 | NP_083689.2 | 2, 3 | 457 | |||
GenBank mRNA | AK017472.1 | - | 2, 3 | 426 | ||
AK032807.1 | - | 2, 3 | 458 | |||
AK048519.1 | - | 2, 3 | 426 | |||
AK053895.1 | - | 2, 3 | 429 | |||
AK164017.1 | - | 2, 3 | 425 | |||
BC098483.1 | - | 2, 3 | 412 | 30022418 | ||
BC132153.1 | - | 2, 3 | 311 | 40130430 | ||
BC132497.1 | - | 2, 3 | 311 | 40130774 |