Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029478.3 | NP_083754.2 | 3, 4 | 323 | ||
GenBank mRNA | AB047550.1 | - | 3, 4 | 335 | ||
AB076609.1 | - | 2, 3 | 214 | |||
AK016312.1 | - | 3, 4 | 340 | |||
AK043091.1 | - | 3, 4 | 324 | |||
AK049559.1 | - | 3, 4 | 316 | |||
AK050796.1 | - | 2, 3 | 192 | |||
AK077443.1 | - | 3, 4 | 323 | |||
AK159468.1 | - | 3, 4 | 338 | |||
AK167173.1 | - | 3, 4 | 331 | |||
AK167202.1 | - | 3, 4 | 331 | |||
AX775963.1 | - | 3, 4 | 293 | |||
BC004013.1 | - | 3, 4 | 317 | 3493738 |