Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029492.4 | NP_083768.4 | 3 | 242 | ||
GenBank mRNA | AK017158.1 | - | 3 | 243 | ||
AK031184.1 | - | 3 | 201 | |||
AK049270.1 | - | 3 | 222 | |||
AK139216.1 | - | 3 | 201 | |||
AK140182.1 | - | 3 | 203 | |||
AK151840.1 | - | 3 | 257 | |||
AK163174.1 | - | 3 | 240 | |||
AK170017.1 | - | 3 | 260 | |||
AK170139.1 | - | 3 | 183 | |||
AY668951.1 | - | 3 | 160 | |||
BC019536.1 | - | 3 | 259 | 4039734 |