Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam131b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113327.2 | NP_001106798.1 | 4 | 434 | ||
NM_001286584.1 | NP_001273513.1 | 5 | 410 | |||
NM_029528.5 | NP_083804.3 | 5 | 470 | |||
GenBank mRNA | AK018206.1 | - | 5 | 409 | ||
AK018325.1 | - | 5 | 416 | |||
AK047763.1 | - | 5 | 396 | |||
AK158869.1 | - | 5 | 363 | |||
AK163654.1 | - | 4 | 355 | |||
BC115595.1 | - | 4 | 200 | |||
BC115596.1 | - | 4 | 200 | |||
BY260739.1 | - | 4 | 435 |