Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029530.2 | NP_083806.2 | 4, 5 | 868 | ||
GenBank mRNA | AK014127.1 | - | 2, 3 | 310 | ||
AK018222.1 | - | 4, 5 | 567 | |||
AK045946.1 | - | 3, 4 | 920 | |||
BC004791.1 | - | 2, 3 | 144 | 3588577 |