Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029555.2 | NP_083831.1 | 8 | 834 | ||
GenBank mRNA | AK002661.1 | - | 8 | 832 | ||
AY279096.1 | - | 8 | 747 | |||
BC038151.1 | - | 8 | 761 | |||
BC051507.1 | - | 8 | 875 | |||
BC058163.1 | - | 8 | 767 | 5100572 |